Product Details
- SNP ID
-
rs113107098
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
6
- Location
-
Chr.1:19251605 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CACAGAGCTGGGATAGCGCGCGAGG[C/G]CCTCGGTCGGACGGGACGCTCGGGA
- Phenotype
-
MIM: 616846
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
EMC1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs141918623] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- EMC1
- Gene Name
- ER membrane protein complex subunit 1
- Gene
- MRTO4
- Gene Name
- MRT4 homolog, ribosome maturation factor
View Full Product Details