Product Details
- SNP ID
-
rs61745399
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:144115649 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACCCTGCCGGGCCTGCTCAGCTGCC[C/T]GGAGAGCCGCCGCCGCTGGAGGAGA
- Phenotype
-
MIM: 610210
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MAF1
PubMed Links
Gene Details
- Gene
- MAF1
- Gene Name
- MAF1 homolog, negative regulator of RNA polymerase III
There are no transcripts associated with this gene.
- Gene
- WDR97
- Gene Name
- WD repeat domain 97
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001316309.1 |
4439 |
Silent Mutation |
CCC,CCT |
P1462P |
NP_001303238.1 |
XM_017013385.1 |
4439 |
Silent Mutation |
CCC,CCT |
P1505P |
XP_016868874.1 |
XM_017013386.1 |
4439 |
Silent Mutation |
CCC,CCT |
P1504P |
XP_016868875.1 |
XM_017013387.1 |
4439 |
Silent Mutation |
CCC,CCT |
P1490P |
XP_016868876.1 |
XM_017013388.1 |
4439 |
Silent Mutation |
CCC,CCT |
P1488P |
XP_016868877.1 |
XM_017013389.1 |
4439 |
Silent Mutation |
CCC,CCT |
P1479P |
XP_016868878.1 |
XM_017013390.1 |
4439 |
Silent Mutation |
CCC,CCT |
P1473P |
XP_016868879.1 |
XM_017013391.1 |
4439 |
Silent Mutation |
CCC,CCT |
P1463P |
XP_016868880.1 |
XM_017013392.1 |
4439 |
Silent Mutation |
CCC,CCT |
P1462P |
XP_016868881.1 |
XM_017013393.1 |
4439 |
Silent Mutation |
CCC,CCT |
P1451P |
XP_016868882.1 |
XM_017013394.1 |
4439 |
Silent Mutation |
CCC,CCT |
P1440P |
XP_016868883.1 |
View Full Product Details