Product Details
- SNP ID
-
rs76291294
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:74091144 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCATATCTCTAGACTTCCAGGTGAG[C/T]GTTCCAGTTTATGTCCTTTGAGATT
- Phenotype
-
MIM: 603178
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ALDH6A1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76957959] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ALDH6A1
- Gene Name
- aldehyde dehydrogenase 6 family member A1
There are no transcripts associated with this gene.
- Gene
- LIN52
- Gene Name
- lin-52 DREAM MuvB core complex component
View Full Product Details