Product Details
- SNP ID
-
rs35374376
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:150065586 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTTTGAGTCAAAGAGGAGGAACTTC[C/T]GTCCATCGAAGCTGAACTGCCAAGA
- Phenotype
-
MIM: 605699
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ULBP3
PubMed Links
Gene Details
- Gene
- ULBP3
- Gene Name
- UL16 binding protein 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_024518.2 |
521 |
Missense Mutation |
|
|
NP_078794.1 |
View Full Product Details