Product Details
- SNP ID
-
rs34315986
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
21
- Location
-
Chr.1:28259865 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGCGCACCATGATCGTGGCGGACTC[C/G]GAGTGCCGCGCAGAGCTCAAGGACT
- Phenotype
-
MIM: 607767
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SESN2
PubMed Links
Gene Details
- Gene
- SESN2
- Gene Name
- sestrin 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_031459.4 |
414 |
Silent Mutation |
TCC,TCG |
S6S |
NP_113647.1 |
View Full Product Details