Product Details
- SNP ID
-
rs61733899
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:71549119 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGTCCAGCTGCTGCAAGCCCTGCTG[C/T]TCCCAGTCCAGATGCTGTGTCCCTG
- Phenotype
-
MIM: 148021
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
KRTAP5-8
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2664] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- KRTAP5-8
- Gene Name
- keratin associated protein 5-8
There are no transcripts associated with this gene.
- Gene
- KRTAP5-9
- Gene Name
- keratin associated protein 5-9
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005553.3 |
700 |
Silent Mutation |
TGC,TGT |
C154C |
NP_005544.4 |
View Full Product Details