Product Details

SNP ID
rs147001251
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:38276902 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAATATTAATTCAAATATTGAAGC[C/T]GAAGAAAAAAATACTGTACAGTCCA
Phenotype
MIM: 312610
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RPGR PubMed Links

Gene Details

Gene
RPGR
Gene Name
retinitis pigmentosa GTPase regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000328.2 Intron NP_000319.1
NM_001034853.1 Intron NP_001030025.1
XM_005272633.2 Intron XP_005272690.1
XM_011543940.2 Intron XP_011542242.1
XM_017029710.1 Intron XP_016885199.1
XM_017029711.1 Intron XP_016885200.1
XM_017029712.1 Intron XP_016885201.1

View Full Product Details