Product Details

SNP ID
rs61737679
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:223765297 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATTTCCCCACCTATTATAAACTC[A/G]TCCAGGATGAAATAAGCCTTTTCAA
Phenotype
MIM: 615781
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AP1S3 PubMed Links

Gene Details

Gene
AP1S3
Gene Name
adaptor related protein complex 1 sigma 3 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039569.1 497 Silent Mutation GAC,GAT D115D NP_001034658.1
XM_011510600.2 497 Intron XP_011508902.1

View Full Product Details