Product Details

SNP ID
rs61746141
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:97320157 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTACGTGCAAAGCTTCCCCAACGCC[C/G]GCTCCTGGGACCTCTGTCGGCCCCG
Phenotype
MIM: 602503
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FRAT1 PubMed Links

Gene Details

Gene
FRAT1
Gene Name
frequently rearranged in advanced T-cell lymphomas 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005479.3 893 Missense Mutation CCG,CGG P235R NP_005470.2

View Full Product Details