Product Details

SNP ID
rs28365946
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:82458529 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGCCGTACGTGGAGTGAGCCTGCG[C/G]TCCGCCCCGCGCGCTCCGATTGGCT
Phenotype
MIM: 605349
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C17orf62 PubMed Links

Gene Details

Gene
C17orf62
Gene Name
chromosome 17 open reading frame 62
There are no transcripts associated with this gene.

Gene
NARF
Gene Name
nuclear prelamin A recognition factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001038618.2 Intron NP_001033707.1
NM_001083608.1 Intron NP_001077077.1
NM_012336.3 Intron NP_036468.1
NM_031968.2 Intron NP_114174.1
XM_005256340.3 Intron XP_005256397.1
XM_006722278.3 Intron XP_006722341.1
XM_011523541.2 Intron XP_011521843.1
XM_011523543.2 Intron XP_011521845.1
XM_011523544.2 Intron XP_011521846.1
XM_017024449.1 Intron XP_016879938.1
XM_017024450.1 Intron XP_016879939.1

View Full Product Details