Product Details

SNP ID
rs61741121
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:88468001 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAAGCAGAAAGTTGAACAGAACGC[G/T]GCTCCCAGCCACACCAAGTTCAGGT
Phenotype
MIM: 611851 MIM: 611977
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MRPL46 PubMed Links

Gene Details

Gene
MRPL46
Gene Name
mitochondrial ribosomal protein L46
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022163.3 421 Intron NP_071446.2
Gene
MRPS11
Gene Name
mitochondrial ribosomal protein S11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321970.1 421 Silent Mutation GCG,GCT A52A NP_001308899.1
NM_001321972.1 421 UTR 5 NP_001308901.1
NM_001321973.1 421 Silent Mutation GCG,GCT A52A NP_001308902.1
NM_001321974.1 421 Silent Mutation GCG,GCT A53A NP_001308903.1
NM_001321976.1 421 UTR 5 NP_001308905.1
NM_022839.4 421 Silent Mutation GCG,GCT A53A NP_073750.2
NM_176805.3 421 Silent Mutation GCG,GCT A53A NP_789775.1

View Full Product Details