Product Details

SNP ID
rs111495538
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:96838010 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCATTTTCTAGATGAGGAGTTGT[C/T]TTCTCCATGGTCTTGAACGCCTGCT
Phenotype
MIM: 610736 MIM: 607804
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANKRD23 PubMed Links

Gene Details

Gene
ANKRD23
Gene Name
ankyrin repeat domain 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144994.7 2486 UTR 3 NP_659431.5
Gene
ANKRD39
Gene Name
ankyrin repeat domain 39
There are no transcripts associated with this gene.

Gene
CNNM3
Gene Name
cyclin and CBS domain divalent metal cation transport mediator 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017623.4 2486 Intron NP_060093.3
NM_199078.2 2486 Intron NP_951060.1
XM_011510957.2 2486 Intron XP_011509259.1
XM_017003800.1 2486 Intron XP_016859289.1

View Full Product Details