Product Details

SNP ID
rs72960626
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:119294867 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTACAGCGGTGCCAAGCAGAGGGG[A/C]GGCAGAGACGGAGGGGCAGCCTCTT
Phenotype
MIM: 610911
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ARHGAP31 PubMed Links

Gene Details

Gene
ARHGAP31
Gene Name
Rho GTPase activating protein 31
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020754.3 495 UTR 5 NP_065805.2
XM_006713714.3 495 UTR 5 XP_006713777.1
XM_017006955.1 495 UTR 5 XP_016862444.1

View Full Product Details