Product Details

SNP ID
rs61741482
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:132552800 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGGTCCCGGGTAATTCTGTACAC[A/G]GCGTTTAGCTTTGGGTCTTTGCTGG
Phenotype
MIM: 606927
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TAAR8 PubMed Links

Gene Details

Gene
TAAR8
Gene Name
trace amine associated receptor 8
There are no transcripts associated with this gene.

View Full Product Details