Product Details

SNP ID
rs73522766
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:69667745 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGACTGTCTTCAGTTTTCCTGCA[C/T]TGCGACTCAAGGATGAGGCGCCTCT
Phenotype
MIM: 607224
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ORAOV1 PubMed Links

Gene Details

Gene
ORAOV1
Gene Name
oral cancer overexpressed 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153451.2 Intron NP_703152.1
XM_006718470.2 Intron XP_006718533.1

View Full Product Details