Product Details

SNP ID
rs73913116
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:9492846 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTTCCAGAGATTACATGGTTGGA[A/G]TTGATCAAAATTTAAATAAAACATT
Phenotype
MIM: 603639
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADAM17 PubMed Links

Gene Details

Gene
ADAM17
Gene Name
ADAM metallopeptidase domain 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003183.5 Intron NP_003174.3
XM_011510375.2 Intron XP_011508677.1
XM_011510376.2 Intron XP_011508678.1
XM_011510377.2 Intron XP_011508679.1
XM_017004785.1 Intron XP_016860274.1
XM_017004786.1 Intron XP_016860275.1
Gene
IAH1
Gene Name
isoamyl acetate-hydrolyzing esterase 1 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039613.2 Intron NP_001034702.1
NM_001320858.1 Intron NP_001307787.1
NM_001320859.1 Intron NP_001307788.1
NM_001320860.1 Intron NP_001307789.1
NM_001320863.1 Intron NP_001307792.1
XM_017003878.1 Intron XP_016859367.1

View Full Product Details