Product Details

SNP ID
rs115549552
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:17435946 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTGCACATCAGCAGCATGGAGAT[G/T]ATGGGGTTCAGGTGAGAATCCTGGC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TMEM221 PubMed Links

Gene Details

Gene
TMEM221
Gene Name
transmembrane protein 221
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190844.1 1493 UTR 3 NP_001177773.1
XM_011527603.2 1493 UTR 3 XP_011525905.1

View Full Product Details