Product Details
- SNP ID
-
rs34604301
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:23452680 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGGTGATCCACTGCAAGCTGTCCTT[C/T]GCATAGTTTTCTACTGCCATCACTT
- Phenotype
-
MIM: 609731
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CST11
PubMed Links
Gene Details
- Gene
- CST11
- Gene Name
- cystatin 11
- Gene
- CSTL1
- Gene Name
- cystatin like 1
There are no transcripts associated with this gene.
View Full Product Details