Product Details
- SNP ID
-
rs35435463
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:74245180 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCCCCTCAGGACAATCTTTACCTC[C/T]TACCAGCTAGAGGAGCTGGAGAAGG
- Phenotype
-
MIM: 142993
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
VSX2
PubMed Links
Gene Details
- Gene
- VSX2
- Gene Name
- visual system homeobox 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_182894.2 |
561 |
Silent Mutation |
TCC,TCT |
S157S |
NP_878314.1 |
View Full Product Details