Product Details

SNP ID
rs146723341
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:42317753 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCTGGATGAGCATAAAGGCTTTG[A/G]GGCTAAAAACCTTGATGGAGATGAA
Phenotype
MIM: 608150 MIM: 610750
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR7851 PubMed Links

Gene Details

Gene
MIR7851
Gene Name
microRNA 7851
There are no transcripts associated with this gene.

Gene
PPHLN1
Gene Name
periphilin 1
There are no transcripts associated with this gene.

Gene
ZCRB1
Gene Name
zinc finger CCHC-type and RNA binding motif containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033114.3 Intron NP_149105.3
XM_017020124.1 Intron XP_016875613.1

View Full Product Details