Product Details

SNP ID
rs3746203
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:15238547 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCAGTCAGCCCCGTAGCCCTCCCC[A/G]TGGCTGACCCCTCATAGCGCTCACC
Phenotype
MIM: 608749
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BRD4 PubMed Links

Gene Details

Gene
BRD4
Gene Name
bromodomain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014299.2 Intron NP_055114.1
NM_058243.2 Intron NP_490597.1
XM_011527854.1 Intron XP_011526156.1
XM_011527855.1 Intron XP_011526157.1
XM_011527856.2 Intron XP_011526158.1
Gene
EPHX3
Gene Name
epoxide hydrolase 3
There are no transcripts associated with this gene.

View Full Product Details