Product Details

SNP ID
rs35117143
Assay Type
Functionally Tested
NCBI dbSNP Submissions
7
Location
Chr.1:89185356 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTAGCCATGTCAAGGATCTTTAA[A/G]AGCCTTAACTGTTCATTTTTAGTGC
Phenotype
MIM: 612466 MIM: 612468
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
GBP4 PubMed Links

Gene Details

Gene
GBP4
Gene Name
guanylate binding protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052941.4 1937 Silent Mutation CTC,CTT L607L NP_443173.2
Gene
GBP7
Gene Name
guanylate binding protein 7
There are no transcripts associated with this gene.

View Full Product Details