Product Details

SNP ID
rs185615734
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:46247070 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAGGGAGAGCTCCAGGATAACCA[C/T]AGCAAGGACAGGTGGCCCCAGCCCA
Phenotype
MIM: 604670 MIM: 170998
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDPF1 PubMed Links

Gene Details

Gene
CDPF1
Gene Name
cysteine rich DPF motif domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207327.4 Intron NP_997210.3
XM_011529960.1 Intron XP_011528262.1
XM_011529962.2 Intron XP_011528264.1
XM_011529963.1 Intron XP_011528265.1
XM_011529964.1 Intron XP_011528266.1
XM_011529965.2 Intron XP_011528267.1
Gene
PKDREJ
Gene Name
polycystin (PKD) family receptor for egg jelly
There are no transcripts associated with this gene.

Gene
PPARA
Gene Name
peroxisome proliferator activated receptor alpha
There are no transcripts associated with this gene.

View Full Product Details