Product Details

SNP ID
rs7253024
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:6362713 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGTGCCTGTTGCTGGCTTTTGAGC[C/T]TCGGTTCACAAGTCAGGATGTGGCC
Phenotype
MIM: 613491 MIM: 613305 MIM: 601119
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACER1 PubMed Links

Gene Details

Gene
ACER1
Gene Name
alkaline ceramidase 1
There are no transcripts associated with this gene.

Gene
ALKBH7
Gene Name
alkB homolog 7
There are no transcripts associated with this gene.

Gene
CLPP
Gene Name
caseinolytic mitochondrial matrix peptidase proteolytic subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006012.2 Intron NP_006003.1

View Full Product Details