Product Details
- SNP ID
-
rs12043009
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
40
- Location
-
Chr.1:153261819 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GCGGCTCCTCCGTGGGTGGCTCCGG[A/G]AGTGGCAAGGGCGTCCCGATCTGCC
- Phenotype
-
MIM: 152445
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
LOR
PubMed Links
Gene Details
- Gene
- LOR
- Gene Name
- loricrin
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000427.2 |
924 |
Silent Mutation |
GGA,GGG |
G290G |
NP_000418.2 |
View Full Product Details