Product Details
- SNP ID
-
rs1142888
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
19
- Location
-
Chr.1:89186407 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTTTCCTCCTCCAACTTCTTCTCCA[T/G]TTGGGCCATGTATTCCTGGAAGCTT
- Phenotype
-
MIM: 612466
- Polymorphism
- T/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
GBP4
PubMed Links
Gene Details
- Gene
- GBP4
- Gene Name
- guanylate binding protein 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_052941.4 |
1749 |
Missense Mutation |
ATG,CTG |
M545L |
NP_443173.2 |
View Full Product Details