Product Details
- SNP ID
-
hCV25967230
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:73407422 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACTTGACAGCTGTTACCTGCCCTAG[C/T]TGACACCTGTGCTCCAGACCCAAAC
- Phenotype
-
MIM: 616316
MIM: 611211
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FAM168A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs113418337] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FAM168A
- Gene Name
- family with sequence similarity 168 member A
- Gene
- RELT
- Gene Name
- RELT tumor necrosis factor receptor
There are no transcripts associated with this gene.
View Full Product Details