Product Details

SNP ID
rs145427230
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:16553137 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATCCACACTTCAATGCTCACAGT[A/C]ATCTCAGATTTAATATTTGATTCTT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FAM49A PubMed Links

Gene Details

Gene
FAM49A
Gene Name
family with sequence similarity 49 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030797.3 Intron NP_110424.1
XM_005262630.3 Intron XP_005262687.1
XM_006712108.3 Intron XP_006712171.1

View Full Product Details