Product Details

SNP ID
rs34987563
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:23561601 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCTCATCAGCCTGAGGGCCATCTC[G/T]CTCCACAAGAGGCATTTTTTCCAGG
Phenotype
MIM: 603534 MIM: 611612
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
AP1G2 PubMed Links

Gene Details

Gene
AP1G2
Gene Name
adaptor related protein complex 1 gamma 2 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282474.1 1740 Silent Mutation NP_001269403.1
NM_001282475.1 1740 Silent Mutation NP_001269404.1
NM_003917.4 1740 Silent Mutation NP_003908.1
XM_005268167.3 1740 Nonsense Mutation XP_005268224.1
XM_005268168.4 1740 Nonsense Mutation XP_005268225.1
XM_005268169.3 1740 Nonsense Mutation XP_005268226.1
XM_005268170.3 1740 Nonsense Mutation XP_005268227.1
XM_005268172.3 1740 Nonsense Mutation XP_005268229.1
XM_005268173.3 1740 Nonsense Mutation XP_005268230.1
XM_005268174.3 1740 Nonsense Mutation XP_005268231.1
XM_005268175.3 1740 Nonsense Mutation XP_005268232.1
XM_005268177.3 1740 Nonsense Mutation XP_005268234.1
XM_005268178.3 1740 Nonsense Mutation XP_005268235.1
XM_005268179.3 1740 Nonsense Mutation XP_005268236.1
XM_005268180.4 1740 Nonsense Mutation XP_005268237.1
XM_005268182.3 1740 Nonsense Mutation XP_005268239.1
XM_006720301.3 1740 Silent Mutation XP_006720364.1
XM_011537283.2 1740 Nonsense Mutation XP_011535585.1
XM_011537286.2 1740 Nonsense Mutation XP_011535588.1
XM_017021740.1 1740 Nonsense Mutation XP_016877229.1
XM_017021741.1 1740 Nonsense Mutation XP_016877230.1
XM_017021742.1 1740 Nonsense Mutation XP_016877231.1
XM_017021743.1 1740 Intron XP_016877232.1
XM_017021744.1 1740 Intron XP_016877233.1
XM_017021745.1 1740 Intron XP_016877234.1
XM_017021746.1 1740 Nonsense Mutation XP_016877235.1
XM_017021747.1 1740 Nonsense Mutation XP_016877236.1
XM_017021748.1 1740 Nonsense Mutation XP_016877237.1
Gene
JPH4
Gene Name
junctophilin 4
There are no transcripts associated with this gene.

Gene
LOC102724814
Gene Name
uncharacterized LOC102724814
There are no transcripts associated with this gene.

Gene
THTPA
Gene Name
thiamine triphosphatase
There are no transcripts associated with this gene.

Gene
ZFHX2
Gene Name
zinc finger homeobox 2
There are no transcripts associated with this gene.

View Full Product Details