Product Details

SNP ID
rs141456097
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:54156745 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGAAGGTCTGGGCCCTGGTCTGCC[A/G]AGGTGGGGTCTTCTTACTCCTTGGG
Phenotype
MIM: 604910
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CNOT3 PubMed Links

Gene Details

Gene
CNOT3
Gene Name
CCR4-NOT transcription complex subunit 3
There are no transcripts associated with this gene.

Gene
LENG1
Gene Name
leukocyte receptor cluster member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024316.2 Intron NP_077292.2
Gene
TMC4
Gene Name
transmembrane channel like 4
There are no transcripts associated with this gene.

View Full Product Details