Product Details

SNP ID
rs114302918
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:27772415 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACTTTAGAAAGATTTAAAAGGTAG[A/C]CAGTCCAACAGTCATGAAATTATGC
Phenotype
MIM: 610059 MIM: 611132
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MRPL33 PubMed Links

Gene Details

Gene
MRPL33
Gene Name
mitochondrial ribosomal protein L33
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004891.3 Intron NP_004882.1
NM_145330.2 Intron NP_663303.1
Gene
RBKS
Gene Name
ribokinase
There are no transcripts associated with this gene.

View Full Product Details