Product Details

SNP ID
hCV25972848
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:68188470 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTACACAGAGCTGTAAGAATACAAA[C/T]AGGTTAGATAACTTGTGGTTATAAA
Phenotype
MIM: 601302
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PPP3R1 PubMed Links

Gene Details

Gene
PPP3R1
Gene Name
protein phosphatase 3 regulatory subunit B, alpha
There are no transcripts associated with this gene.

View Full Product Details