Product Details

SNP ID
rs3760843
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:45785448 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTCTAAGGATCTGAGAGGTCTTGT[A/T]CTGAGACACTAAGATTTCCCTGGCT
Phenotype
MIM: 605377 MIM: 609857
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
DMPK PubMed Links

Gene Details

Gene
DMPK
Gene Name
dystrophia myotonica protein kinase
There are no transcripts associated with this gene.

Gene
DMWD
Gene Name
dystrophia myotonica, WD repeat containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004943.1 Intron NP_004934.1

View Full Product Details