Product Details

SNP ID
rs74090784
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:53504394 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACTTTGGCCCTGTGCCTTTTCCTT[C/G]AGGAGCCCCCCCATGGAACTGCAGC
Phenotype
MIM: 606371 MIM: 600447 MIM: 604643 MIM: 605053
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ATF7 PubMed Links

Gene Details

Gene
ATF7
Gene Name
activating transcription factor 7
There are no transcripts associated with this gene.

Gene
LOC100652999
Gene Name
uncharacterized LOC100652999
There are no transcripts associated with this gene.

Gene
MAP3K12
Gene Name
mitogen-activated protein kinase kinase kinase 12
There are no transcripts associated with this gene.

Gene
NPFF
Gene Name
neuropeptide FF-amide peptide precursor
There are no transcripts associated with this gene.

Gene
TARBP2
Gene Name
TARBP2, RISC loading complex RNA binding subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004178.4 Intron NP_004169.3
NM_134323.1 Intron NP_599150.1
NM_134324.2 Intron NP_599151.2
XM_005269114.1 Intron XP_005269171.1
XM_005269115.2 Intron XP_005269172.1
XM_005269117.1 Intron XP_005269174.1
XM_005269120.4 Intron XP_005269177.1
XM_005269122.3 Intron XP_005269179.1
XM_006719581.1 Intron XP_006719644.1
XM_011538712.2 Intron XP_011537014.1
XM_017019910.1 Intron XP_016875399.1
XM_017019911.1 Intron XP_016875400.1

View Full Product Details