Product Details
- SNP ID
-
rs9895154
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:44190581 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTACAAGACCCAGCCCGCACACTGC[A/G]TTCTCTGAACATTACCGACAACTGT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ASB16-AS1
PubMed Links
Gene Details
- Gene
- ASB16-AS1
- Gene Name
- ASB16 antisense RNA 1
There are no transcripts associated with this gene.
- Gene
- ATXN7L3
- Gene Name
- ataxin 7 like 3
There are no transcripts associated with this gene.
- Gene
- LOC101926967
- Gene Name
- uncharacterized LOC101926967
There are no transcripts associated with this gene.
- Gene
- TMUB2
- Gene Name
- transmembrane and ubiquitin like domain containing 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001076674.1 |
1273 |
Missense Mutation |
CAT,CGT |
H228R |
NP_001070142.1 |
NM_024107.2 |
1273 |
Missense Mutation |
CAT,CGT |
H208R |
NP_077012.2 |
NM_177441.2 |
1273 |
Missense Mutation |
CAT,CGT |
H208R |
NP_803190.2 |
XM_011525198.2 |
1273 |
Missense Mutation |
CAT,CGT |
H228R |
XP_011523500.1 |
XM_011525200.2 |
1273 |
Missense Mutation |
CAT,CGT |
H281R |
XP_011523502.1 |
XM_011525201.2 |
1273 |
Missense Mutation |
CAT,CGT |
H228R |
XP_011523503.1 |
XM_011525203.2 |
1273 |
Missense Mutation |
CAT,CGT |
H219R |
XP_011523505.1 |
XM_011525204.2 |
1273 |
Missense Mutation |
CAT,CGT |
H219R |
XP_011523506.1 |
XM_011525205.2 |
1273 |
Missense Mutation |
CAT,CGT |
H208R |
XP_011523507.1 |
XM_011525206.2 |
1273 |
Missense Mutation |
CAT,CGT |
H208R |
XP_011523508.1 |
XM_011525207.2 |
1273 |
Missense Mutation |
CAT,CGT |
H208R |
XP_011523509.1 |
XM_011525208.2 |
1273 |
Missense Mutation |
CAT,CGT |
H208R |
XP_011523510.1 |
XM_011525209.2 |
1273 |
Missense Mutation |
CAT,CGT |
H208R |
XP_011523511.1 |
XM_011525210.2 |
1273 |
Missense Mutation |
CAT,CGT |
H171R |
XP_011523512.1 |
XM_011525211.2 |
1273 |
UTR 3 |
|
|
XP_011523513.1 |
XM_017025047.1 |
1273 |
Missense Mutation |
CAT,CGT |
H219R |
XP_016880536.1 |
XM_017025048.1 |
1273 |
Missense Mutation |
CAT,CGT |
H208R |
XP_016880537.1 |
XM_017025049.1 |
1273 |
Missense Mutation |
CAT,CGT |
H208R |
XP_016880538.1 |
XM_017025050.1 |
1273 |
Missense Mutation |
CAT,CGT |
H208R |
XP_016880539.1 |
XM_017025051.1 |
1273 |
Missense Mutation |
CAT,CGT |
H208R |
XP_016880540.1 |
XM_017025052.1 |
1273 |
Missense Mutation |
CAT,CGT |
H182R |
XP_016880541.1 |
XM_017025053.1 |
1273 |
Missense Mutation |
CAT,CGT |
H182R |
XP_016880542.1 |
XM_017025054.1 |
1273 |
Missense Mutation |
CAT,CGT |
H171R |
XP_016880543.1 |
XM_017025055.1 |
1273 |
Missense Mutation |
CAT,CGT |
H171R |
XP_016880544.1 |
XM_017025056.1 |
1273 |
UTR 3 |
|
|
XP_016880545.1 |
View Full Product Details