Product Details

SNP ID
rs61738512
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:54631736 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGGTATCGCTGTTACTATGGTAGC[C/G]ACACTGCAGGCCGCTCAGAGAGCAG
Phenotype
MIM: 604811
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LILRB1 PubMed Links

Gene Details

Gene
LILRB1
Gene Name
leukocyte immunoglobulin like receptor B1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001081637.2 578 Intron NP_001075106.2
NM_001081638.3 578 Missense Mutation CAC,GAC H103D NP_001075107.2
NM_001081639.3 578 Missense Mutation CAC,GAC H103D NP_001075108.2
NM_001278398.2 578 Intron NP_001265327.2
NM_001278399.2 578 Missense Mutation CAC,GAC H103D NP_001265328.2
NM_006669.6 578 Missense Mutation CAC,GAC H103D NP_006660.4
XM_011526331.2 578 Intron XP_011524633.1
XM_011526332.2 578 Intron XP_011524634.1
XM_011526335.2 578 Intron XP_011524637.1
XM_011526336.2 578 Intron XP_011524638.1
XM_011526338.2 578 Intron XP_011524640.1
XM_017026182.1 578 Intron XP_016881671.1
XM_017026183.1 578 Intron XP_016881672.1
XM_017026184.1 578 Intron XP_016881673.1
XM_017026185.1 578 Intron XP_016881674.1
XM_017026186.1 578 Intron XP_016881675.1
XM_017026187.1 578 Missense Mutation CAC,GAC H120D XP_016881676.1
XM_017026188.1 578 Missense Mutation CAC,GAC H120D XP_016881677.1
XM_017026189.1 578 Missense Mutation CAC,GAC H120D XP_016881678.1
XM_017026190.1 578 Missense Mutation CAC,GAC H120D XP_016881679.1
XM_017026191.1 578 Intron XP_016881680.1
XM_017026192.1 578 Intron XP_016881681.1

View Full Product Details