Product Details

SNP ID
rs7949943
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:125677765 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGTTCCCCATTTCCCACCTGAAGT[C/G]AATGACTGGCACCCATCCAAACATG
Phenotype
MIM: 102525 MIM: 603078
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ACRV1 PubMed Links

Gene Details

Gene
ACRV1
Gene Name
acrosomal vesicle protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001612.5 Intron NP_001603.1
NM_020069.4 Intron NP_064454.1
NM_020107.4 Intron NP_064492.1
NM_020108.4 Intron NP_064493.1
Gene
CHEK1
Gene Name
checkpoint kinase 1
There are no transcripts associated with this gene.

View Full Product Details