Product Details

SNP ID
rs12291603
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:112230111 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTGGGGACAGATCTAATAATTTA[C/T]GTTGCCAACTTGTGCTTGTATGTTG
Phenotype
MIM: 612719
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PTS PubMed Links

Gene Details

Gene
PTS
Gene Name
6-pyruvoyltetrahydropterin synthase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000317.2 Intron NP_000308.1

View Full Product Details