Product Details

SNP ID
rs3741843
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:10938833 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCAGCACCAAAACCACCTTTTTAAC[C/T]CTCCACTTTAGGTAGAGAAAAATAG
Phenotype
MIM: 168730 MIM: 168790 MIM: 604790
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PRH1 PubMed Links

Gene Details

Gene
PRH1
Gene Name
proline rich protein HaeIII subfamily 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291314.1 375 Intron NP_001278243.1
NM_001291315.1 375 Intron NP_001278244.1
Gene
PRH1-PRR4
Gene Name
PRH1-PRR4 readthrough
There are no transcripts associated with this gene.

Gene
PRH1-TAS2R14
Gene Name
PRH1-TAS2R14 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001316893.1 375 Intron NP_001303822.1
Gene
PRH2
Gene Name
proline rich protein HaeIII subfamily 2
There are no transcripts associated with this gene.

Gene
TAS2R14
Gene Name
taste 2 receptor member 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_023922.1 375 Silent Mutation AGA,AGG R125R NP_076411.1

View Full Product Details