Product Details

SNP ID
rs36182532
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:7088679 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACGGAACTGGTACTGAGGCTGCAG[A/G]TTCTGGATGATGGTGAACTCGTCTA
Phenotype
MIM: 613785 MIM: 608974
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C1R PubMed Links

Gene Details

Gene
C1R
Gene Name
complement C1r subcomponent
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001733.4 1102 Silent Mutation AAC,AAT N323N NP_001724.3
XM_017019915.1 1102 Silent Mutation AAC,AAT N337N XP_016875404.1
Gene
C1RL
Gene Name
complement C1r subcomponent like
There are no transcripts associated with this gene.

View Full Product Details