Product Details
- SNP ID
-
rs7934670
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:5788078 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ACATAGGTGAGGACTATGGCACAGA[A/T]GTGGGCAGTGCAGGTGCTGAAGGCC
- Phenotype
-
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
OR52N1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs200275481,rs368530180] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OR52N1
- Gene Name
- olfactory receptor family 52 subfamily N member 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001001913.1 |
739 |
Missense Mutation |
ATC,TTC |
I247F |
NP_001001913.1 |
- Gene
- OR52N5
- Gene Name
- olfactory receptor family 52 subfamily N member 5
There are no transcripts associated with this gene.
View Full Product Details