Product Details

SNP ID
rs13439524
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:33500300 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATTACATAAGGATAATGAGGCCCA[A/G]CTGAAACCAAGTTTCAGTCTGCCTT
Phenotype
MIM: 614426
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MAK16 PubMed Links

Gene Details

Gene
MAK16
Gene Name
MAK16 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032509.3 2614 UTR 3 NP_115898.2
Gene
TTI2
Gene Name
TELO2 interacting protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001102401.2 2614 Intron NP_001095871.1
NM_001265581.1 2614 Intron NP_001252510.1
NM_025115.3 2614 Intron NP_079391.2
XM_011544665.2 2614 Intron XP_011542967.1

View Full Product Details