Product Details

SNP ID
rs41264915
Assay Type
Functionally tested
NCBI dbSNP Submissions
37
Location
Chr.1:155197995 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTATCCCTCCCAGGCCCCCCGTCCC[A/G]GTAGCCCTGTCTGATAGCACCTGCC
Phenotype
MIM: 158340 MIM: 188062
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR92B PubMed Links

Gene Details

Gene
MIR92B
Gene Name
microRNA 92b
There are no transcripts associated with this gene.

Gene
MUC1
Gene Name
mucin 1, cell surface associated
There are no transcripts associated with this gene.

Gene
THBS3
Gene Name
thrombospondin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001252607.1 Intron NP_001239536.1
NM_001252608.1 Intron NP_001239537.1
NM_007112.4 Intron NP_009043.1
XM_006711498.3 Intron XP_006711561.1
XM_011509926.2 Intron XP_011508228.1
XM_011509927.2 Intron XP_011508229.1
XM_011509928.2 Intron XP_011508230.1
XM_011509929.1 Intron XP_011508231.1
XM_011509930.2 Intron XP_011508232.1
XM_011509931.1 Intron XP_011508233.1
XM_011509932.2 Intron XP_011508234.1
XM_011509933.1 Intron XP_011508235.1
XM_011509934.1 Intron XP_011508236.1
XM_011509935.1 Intron XP_011508237.1
XM_011509936.1 Intron XP_011508238.1
XM_017002206.1 Intron XP_016857695.1

View Full Product Details