Product Details

SNP ID
rs185392081
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:17137387 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTTCTCTGATTTACAATAAAGTC[A/G]TAATGTACAAATCTGGAGAGCCAGA
Phenotype
MIM: 602997 MIM: 602478
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CUBN PubMed Links

Gene Details

Gene
CUBN
Gene Name
cubilin
There are no transcripts associated with this gene.

Gene
TRDMT1
Gene Name
tRNA aspartic acid methyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321006.1 13266 UTR 3 NP_001307935.1
NM_001321007.1 13266 UTR 3 NP_001307936.1
NM_004412.6 13266 UTR 3 NP_004403.1
XM_005252372.4 13266 Intron XP_005252429.1
XM_005252374.4 13266 UTR 3 XP_005252431.1
XM_005252375.4 13266 Intron XP_005252432.1
XM_005252378.3 13266 Intron XP_005252435.1
XM_006717388.3 13266 Intron XP_006717451.1
XM_011519343.2 13266 Intron XP_011517645.1
XM_011519344.2 13266 Intron XP_011517646.1
XM_011519345.2 13266 Intron XP_011517647.1
XM_017015800.1 13266 Intron XP_016871289.1
XM_017015801.1 13266 Intron XP_016871290.1
XM_017015802.1 13266 Intron XP_016871291.1
XM_017015803.1 13266 Intron XP_016871292.1
XM_017015804.1 13266 Intron XP_016871293.1
XM_017015805.1 13266 Intron XP_016871294.1
XM_017015806.1 13266 Intron XP_016871295.1
XM_017015807.1 13266 Intron XP_016871296.1
XM_017015808.1 13266 Intron XP_016871297.1
XM_017015809.1 13266 Intron XP_016871298.1
XM_017015810.1 13266 Intron XP_016871299.1
XM_017015811.1 13266 Intron XP_016871300.1
XM_017015812.1 13266 Intron XP_016871301.1

View Full Product Details