Product Details

SNP ID
rs180720258
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:7737381 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCAATAAAAAGACTCTTTATCAG[A/T]TAAGAAAGGAAACAGATTAGCTGAC
Phenotype
MIM: 606677
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CLEC4C PubMed Links

Gene Details

Gene
CLEC4C
Gene Name
C-type lectin domain family 4 member C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_130441.2 Intron NP_569708.1
NM_203503.1 Intron NP_987099.1
XM_017018940.1 Intron XP_016874429.1

View Full Product Details