Product Details
- SNP ID
-
rs41307763
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
34
- Location
-
Chr.1:16398835 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGAGTTGTGTTCCCTGGGGAGAGGA[A/G]TAGGGCAGAAGGGTGGGAGACATGT
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
SPATA21
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs147405482] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SPATA21
- Gene Name
- spermatogenesis associated 21
- Gene
- SZRD1
- Gene Name
- SUZ RNA binding domain containing 1
View Full Product Details