Product Details

SNP ID
rs61743318
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:41190465 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGCAGCACACCCTGTTGCCAGCC[A/G]ACCTGTGGTGGGTCCAGCTGCTGTA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KRTAP4-1 PubMed Links

Gene Details

Gene
KRTAP4-1
Gene Name
keratin associated protein 4-1
There are no transcripts associated with this gene.

Gene
KRTAP9-1
Gene Name
keratin associated protein 9-1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190460.1 579 Silent Mutation CCA,CCG P193P NP_001177389.1
XM_005257633.1 579 Intron XP_005257690.1

View Full Product Details