Product Details
- SNP ID
-
rs11769461
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:99279644 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGGGCATGGCGGCCTCACTGAGTGC[A/C]GCCAAGCGCAAGTTGGAAGGGGAGC
- Phenotype
-
MIM: 608580
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
MYH16
PubMed Links
Gene Details
- Gene
- MYH16
- Gene Name
- myosin heavy chain 16 pseudogene
There are no transcripts associated with this gene.
View Full Product Details