Product Details

SNP ID
rs73921005
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:63243908 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGCCGTGAGCAAGGCCCTGCTGGG[C/T]GTGAGGTCCCTTCCAAGTGGGGAGG
Phenotype
MIM: 605737 MIM: 612873
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BIRC7 PubMed Links

Gene Details

Gene
BIRC7
Gene Name
baculoviral IAP repeat containing 7
There are no transcripts associated with this gene.

Gene
LOC107985425
Gene Name
uncharacterized LOC107985425
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017028201.1 Intron XP_016883690.1
Gene
MIR3196
Gene Name
microRNA 3196
There are no transcripts associated with this gene.

Gene
NKAIN4
Gene Name
Na+/K+ transporting ATPase interacting 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152864.3 Intron NP_690603.3
XM_005260192.1 Intron XP_005260249.1
XM_005260194.3 Intron XP_005260251.1
XM_011528527.1 Intron XP_011526829.1
XM_011528528.2 Intron XP_011526830.1
XM_011528529.2 Intron XP_011526831.1
XM_017027636.1 Intron XP_016883125.1
XM_017027637.1 Intron XP_016883126.1
XM_017027638.1 Intron XP_016883127.1
XM_017027639.1 Intron XP_016883128.1

View Full Product Details