Product Details
- SNP ID
-
rs192841031
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:74550066 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCTCTATCATTTGCTTTTCCAAGT[C/T]TCCCACCAATCCAAATGATCCCCCA
- Phenotype
-
MIM: 602919
MIM: 607163
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
DOK1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79033768] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DOK1
- Gene Name
- docking protein 1
- Gene
- LOXL3
- Gene Name
- lysyl oxidase like 3
- Gene
- M1AP
- Gene Name
- meiosis 1 associated protein
There are no transcripts associated with this gene.
View Full Product Details